4-Brom-2-chlorbenzolsulfonylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Brom-2-chlorbenzolsulfonsäurechlorid, 4-Brom-2-chlorbenzolsulfochlorid
M-Clarity™ Qualitätsstufe = MQ100
558729-5G
SIAL558729-5GEA
43.7
EUR
SIAL558729-5G
4-Brom-2-chlorbenzolsulfonylchlorid, Sigma-Aldrich®
4-Brom-2-chlorbenzolsulfonylchlorid
Formel:
C₆H₃BrCl₂O₂S Molecular Weight: 289,96 g/mol Schmelzpunkt: 76…81 °C |
MDL:
MFCD03094630 CAS-Nummer: 351003-52-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...