4-Brom-2,6-dichlorbenzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brom-2,6-dichlorbenzolsulfochlorid, 4-Brom-2,6-dichlorbenzolsulfonsäurechlorid

M-Clarity™ Qualitätsstufe = MQ100

558745-5G
SIAL558745-5GEA 116 EUR
SIAL558745-5G
4-Brom-2,6-dichlorbenzolsulfonylchlorid, Sigma-Aldrich®
4-Brom-2,6-dichlorbenzolsulfonylchlorid
Formel: C₆H₂BrCl₃O₂S
Molecular Weight: 324,41 g/mol
MDL: MFCD03094651
CAS-Nummer: 351003-54-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR