(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-3-(3-methoxyphenyl)propanoic acid

55894-500MG-F
SIAL55894-500MG-FEA 340 EUR
SIAL55894-500MG-F
(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(3-methoxyphenyl)propionsäure
Formel: C₁₅H₂₁NO₅
Molecular Weight: 295,3309 g/mol
MDL: MFCD03427899
CAS-Nummer: 499995-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR