2-Phenyl-1,3-propandiol, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
559296-5G
SIAL559296-5GEA
157
EUR
SIAL559296-5G
2-Phenyl-1,3-propandiol, Sigma-Aldrich®
2-Phenyl-1,3-propandiol
Formel:
HOH₂CCH(C₆H₅)CH₂OH |
MDL:
MFCD00236056 CAS-Nummer: 1570-95-2 EINECS: 411-810-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...