2'-Chlor-5'-(trifluormethyl)acetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

559512-1G
SIAL559512-1GEA 66 EUR
SIAL559512-1G
2'-Chlor-5'-(trifluormethyl)acetophenon, Sigma-Aldrich®
2'-Chlor-5'-(trifluormethyl)acetophenon
Formel: C₉H₆ClF₃O
Molecular Weight: 222,59 g/mol
MDL: MFCD00045092
CAS-Nummer: 71648-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR