2,6-Dinitro-4-(trifluormethyl)anilin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Amino-3,5-dinitro-α,α,α-trifluortoluol, 2,6-Dinitro-α,α,α-trifluor-p-toluidin
M-Clarity™ Qualitätsstufe = MQ100
559520-5G
SIAL559520-5GEA
82.2
EUR
SIAL559520-5G
2,6-Dinitro-4-(trifluormethyl)anilin, Sigma-Aldrich®
2,6-Dinitro-4-(trifluormethyl)anilin
Formel:
C₇H₄F₃N₃O₄ Molecular Weight: 251,12 g/mol |
MDL:
MFCD00024260 CAS-Nummer: 445-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...