3,5-Difluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Difluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

559563-1G
SIAL559563-1GEA 26.6 EUR
SIAL559563-1G
3,5-Difluorbenzolsulfonamid, Sigma-Aldrich®
3,5-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD02091379
CAS-Nummer: 140480-89-3
UN: 3144
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR