3,4-Difluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Difluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

559571-1G
SIAL559571-1GEA 46.8 EUR
SIAL559571-1G
3,4-Difluorbenzolsulfonamid, Sigma-Aldrich®
3,4-Difluorbenzolsulfonamid
Formel: C₆H₅F₂NO₂S
Molecular Weight: 193,17 g/mol
MDL: MFCD00728800
CAS-Nummer: 108966-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR