3-Fluor-4-methylbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

559644-1G
SIAL559644-1GEA 50 EUR
SIAL559644-1G
3-Fluor-4-methylbenzolsulfonamid, Sigma-Aldrich®
3-Fluor-4-methylbenzolsulfonamid
Formel: C₇H₈FNO₂S
MDL: MFCD02710419
CAS-Nummer: 329909-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR