4-Brom-3-fluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brom-3-fluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

559687-1G
SIAL559687-1GEA 43.2 EUR
SIAL559687-1G
4-Brom-3-fluorbenzolsulfonamid, Sigma-Aldrich®
4-Brom-3-fluorbenzolsulfonamid
Formel: C₆H₅BrFNO₂S
Molecular Weight: 254,08 g/mol
MDL: MFCD03094292
CAS-Nummer: 263349-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR