4-Brom-3-(trifluormethyl)benzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brom-3-(trifluormethyl)benzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

559725-1G
SIAL559725-1GEA 33.5 EUR
SIAL559725-1G
4-Brom-3-(trifluormethyl)benzolsulfonamid, Sigma-Aldrich®
4-Brom-3-(trifluormethyl)benzolsulfonamid
Formel: C₇H₅BrF₃NO₂S
Molecular Weight: 304,09 g/mol
MDL: MFCD03094285
CAS-Nummer: 351003-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR