4-Nitro-2,3-lutidin-N-oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3-Dimethyl-4-nitropyridine 1-oxide, 2,3-Dimethyl-4-nitropyridine N-oxide

M-Clarity™ Qualitätsstufe = MQ100

559849-25G
SIAL559849-25GEA 82.4 EUR
SIAL559849-25G
4-Nitro-2,3-lutidin-N-oxid, Sigma-Aldrich®
4-Nitro-2,3-lutidin-N-oxid
Formel: C₇H₈N₂O₂
Molecular Weight: 168,15 g/mol
MDL: MFCD00065172
CAS-Nummer: 37699-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR