2,4,5-Trifluor-4-hydroxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

559962-1G
SIAL559962-1GEA 28.1 EUR
SIAL559962-1G
2,4,5-Trifluor-4-hydroxybenzoesäure, Sigma-Aldrich®
2,4,5-Trifluor-4-hydroxybenzoesäure
Formel: C₇H₃F₃O₃
Molecular Weight: 192,09 g/mol
MDL: MFCD00800378
CAS-Nummer: 116751-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR