3-Brom-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

560162-5G
SIAL560162-5GEA 88.3 EUR
SIAL560162-5G
3-Brom-o-toluylsäure, Sigma-Aldrich®
3-Brom-o-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD00270097
CAS-Nummer: 76006-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR