3-Benzylanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

560243-1G
SIAL560243-1GEA 57.8 EUR
SIAL560243-1G
3-Benzylanilin, Sigma-Aldrich®
3-Benzylanilin
Formel: C₁₃H₁₃N
Molecular Weight: 183,25 g/mol
MDL: MFCD01310782
CAS-Nummer: 61424-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR