2,6-Dichlorphenethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(2-Aminoethyl)-1,3-dichlorobenzene, 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2,6-Dichlorophenyl)ethylamine, 2,6-Dichlorophenethylamine

M-Clarity™ Qualitätsstufe = MQ100

560413-5G
SIAL560413-5GEA 191 EUR
SIAL560413-5G
2,6-Dichlorphenethylamin, Sigma-Aldrich®
2,6-Dichlorphenethylamin
Formel: C₈H₉Cl₂N
Molecular Weight: 190.07 g/mol
MDL: MFCD00060612
CAS-Nummer: 14573-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR