2-(4-Morpholino)ethylisothiocyanat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

560472-2G
SIAL560472-2GEA 67.9 EUR
SIAL560472-2G
2-(4-Morpholino)ethylisothiocyanat, Sigma-Aldrich®
2-(4-Morpholino)ethylisothiocyanat
Formel: C₇H₁₂N₂OS
Molecular Weight: 172,25 g/mol
MDL: MFCD00041221
CAS-Nummer: 63224-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR