3-(4-Fluorphenyl)propionsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
560502-1G
SIAL560502-1GEA
54.2
EUR
SIAL560502-1G
3-(4-Fluorphenyl)propionsäure, Sigma-Aldrich®
3-(4-Fluorphenyl)propionsäure
Formel:
C₉H₉FO₂ Molecular Weight: 168.17 g/mol |
MDL:
MFCD00060327 CAS-Nummer: 459-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...