Diethyl(2-oxobutyl)phosphonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

561053-5ML
SIAL561053-5MLEA 176 EUR
SIAL561053-5ML
Diethyl(2-oxobutyl)phosphonat, Sigma-Aldrich®
Diethyl(2-oxobutyl)phosphonat
Formel: C₈H₁₇O₄P
CAS-Nummer: 1067-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR