4-(4-Hydroxyphenyl)cyclohexanon, Sigma-Aldrich®

Lieferant: Merck

56117-25G-F
SIAL56117-25G-FEA 107 EUR
SIAL56117-25G-F
4-(4-Hydroxyphenyl)cyclohexanon, Sigma-Aldrich®
4-(4-Hydroxyphenyl)cyclohexanone
Formel: C₁₂H₁₄O₂
Molecular Weight: 190,24 g/mol
MDL: MFCD00210693
CAS-Nummer: 105640-07-1
EINECS: 423-920-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR