1,2-Benzisothiazol-3(2H)-on, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

561487-25G 561487-5G
SIAL561487-25GEA 189 EUR
SIAL561487-25G SIAL561487-5G
1,2-Benzisothiazol-3(2H)-on, Sigma-Aldrich®
1,2-Benzisothiazol-3(2H)-on
Formel: C₇H₅NOS
MDL: MFCD00044001
CAS-Nummer: 2634-33-5
EINECS: 220-120-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR