Bis(tetramethylcyclopentadienyl)mangan(II), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(tetramethylcyclopentadienyl)manganese

562424-1G
SIAL562424-1GEA 181 EUR
SIAL562424-1G
Bis(tetramethylcyclopentadienyl)mangan(II), Sigma-Aldrich®
Bis(tetramethylcyclopentadienyl)mangan(II)
Formel: C₁₈H₂₆Mn
Molecular Weight: 297,34 g/mol
MDL: MFCD01862454
CAS-Nummer: 101932-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR