Bis(tetramethylcyclopentadienyl)chrom(II), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(tetramethylcyclopentadienyl)chromium

M-Clarity™ Qualitätsstufe = MQ100

562432-1G
SIAL562432-1GEA 99 EUR
SIAL562432-1G
Bis(tetramethylcyclopentadienyl)chrom(II), Sigma-Aldrich®
Bis(tetramethylcyclopentadienyl)chrom(II)
Formel: C₁₈H₂₆Cr
Molecular Weight: 294,40 g/mol
MDL: MFCD01862451
CAS-Nummer: 82066-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR