Perfluor-1-butansulfonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
562629-25G
562629-5G
SIAL562629-25GEA
204
EUR
SIAL562629-25G
SIAL562629-5G
Perfluor-1-butansulfonsäure, Sigma-Aldrich®
Perfluor-1-butansulfonsäure
Formel:
C₄HF₉O₃S |
MDL:
MFCD01320794 CAS-Nummer: 375-73-5 EINECS: 206-793-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...