(S)-(+)-2-Chlormandelsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2-(2-Chlorophenyl)-2-hydroxyacetic acid

M-Clarity™ Qualitätsstufe = MQ100

562823-500MG
SIAL562823-500MGEA 60.7 EUR
SIAL562823-500MG
(S)-(+)-2-Chlormandelsäure, Sigma-Aldrich®
(S)-(+)-2-Chlormandelsäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD00798437
CAS-Nummer: 52950-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR