6-Brompiperonal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Brom-1,3-benzodioxol-5-carbaldehyd

M-Clarity™ Qualitätsstufe = MQ100

563013-10G
SIAL563013-10GEA 78.5 EUR
SIAL563013-10G
6-Brompiperonal, Sigma-Aldrich®
6-Brompiperonal
Formel: C₈H₅BrO₃
Molecular Weight: 229,03 g/mol
MDL: MFCD00022952
CAS-Nummer: 15930-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR