Lithiumdihydrogenorthophosphat, Sigma-Aldrich®

Lieferant: Merck

563161-25G
SIAL563161-25GEA 105 EUR
SIAL563161-25G
Lithiumdihydrogenorthophosphat, Sigma-Aldrich®
Lithium dihydrogen orthophosphate
Formel: H₂LiO₄P
MDL: MFCD00016188
CAS-Nummer: 13453-80-0
EINECS: 236-633-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR