2-Brom-5-fluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

563498-1G 563498-5G
SIAL563498-1GEA 0 EUR
SIAL563498-1G SIAL563498-5G
2-Brom-5-fluorbenzoesäure, Sigma-Aldrich®
2-Bromo-5-fluorobenzoic acid
Formel: C₇H₄BrFO₂
MDL: MFCD00142874
CAS-Nummer: 394-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR