1-Iod-3-(trifluormethoxy)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

563544-5G
SIAL563544-5GEA 87.2 EUR
SIAL563544-5G
1-Iod-3-(trifluormethoxy)benzol, Sigma-Aldrich®
1-Iod-3-(trifluormethoxy)benzol
Formel: C₇H₄F₃IO
Molecular Weight: 288,01 g/mol
MDL: MFCD01090992
CAS-Nummer: 198206-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR