2-(Trifluormethyl)benzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Trifluormethyl)benzolsulfonsäureamid, α,α,α-Trifluor-o-toluolsulfonamid

M-Clarity™ Qualitätsstufe = MQ100

563552-1G
SIAL563552-1GEA 36.1 EUR
SIAL563552-1G
2-(Trifluormethyl)benzolsulfonamid, Sigma-Aldrich®
2-(Trifluormethyl)benzolsulfonamid
Formel: C₇H₆F₃NO₂S
Molecular Weight: 225,19 g/mol
MDL: MFCD00042420
CAS-Nummer: 1869-24-5
UN: 2811
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR