3-(Trifluormethyl)benzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Trifluormethyl)benzolsulfonsäureamid, α,α,α-Trifluor-m-toluolsulfonamid

M-Clarity™ Qualitätsstufe = MQ100

563560-1G
SIAL563560-1GEA 47 EUR
SIAL563560-1G
3-(Trifluormethyl)benzolsulfonamid, Sigma-Aldrich®
3-(Trifluormethyl)benzolsulfonamid
Formel: C₇H₆F₃NO₂S
Molecular Weight: 225,19 g/mol
MDL: MFCD00042421
CAS-Nummer: 672-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR