4-(Trifluormethyl)benzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: α,α,α-Trifluor-p-toluolsulfonamid, 4-(Trifluormethyl)benzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ200

563579-1G
SIAL563579-1GEA 72 EUR
SIAL563579-1G
4-(Trifluormethyl)benzolsulfonamid, Sigma-Aldrich®
4-(Trifluormethyl)benzolsulfonamid
Formel: C₇H₆F₃NO₂S
Molecular Weight: 225,19 g/mol
MDL: MFCD00159251
CAS-Nummer: 830-43-3
EINECS: 212-596-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR