3-Brombenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brombenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

563609-1G
SIAL563609-1GEA 26 EUR
SIAL563609-1G
3-Brombenzolsulfonamid, Sigma-Aldrich®
3-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
MDL: MFCD00084903
CAS-Nummer: 89599-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR