4-Brom-2-fluorbenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brom-2-fluorbenzolsulfonsäureamid

M-Clarity™ Qualitätsstufe = MQ100

563625-1G
SIAL563625-1GEA 38.5 EUR
SIAL563625-1G
4-Brom-2-fluorbenzolsulfonamid, Sigma-Aldrich®
4-Brom-2-fluorbenzolsulfonamid
Formel: C₆H₅BrFNO₂S
Molecular Weight: 254,08 g/mol
MDL: MFCD03094291
CAS-Nummer: 214210-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR