4-Amino-2-brom-5-pyrimidincarbonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

563846-1G
SIAL563846-1GEA 63 EUR
SIAL563846-1G
4-Amino-2-brom-5-pyrimidincarbonitril, Sigma-Aldrich®
4-Amino-2-brom-5-pyrimidincarbonitril
Formel: C₅H₃BrN₄
Molecular Weight: 199,01 g/mol
MDL: MFCD00173661
CAS-Nummer: 94741-70-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR