1-(Phenylsulfonyl)-3-indolboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1-(Phenylsulphonyl)-1H-indol-3-yl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

563870-1G
SIAL563870-1GEA 77.9 EUR
SIAL563870-1G
1-(Phenylsulfonyl)-3-indolboronsäure, Sigma-Aldrich®
1-(Phenylsulphonyl)-3-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
Molecular Weight: 301,1300 g/mol
MDL: MFCD02681892
CAS-Nummer: 129271-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR