Natrium-1-hydroxy-2,5-dioxo-3-pyrrolidinsulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide

56485-250MG 56485-1G 56485-5G
SIAL56485-250MGEA 104 EUR
SIAL56485-250MG SIAL56485-1G SIAL56485-5G
Natrium-1-hydroxy-2,5-dioxo-3-pyrrolidinsulfonat, Sigma-Aldrich®
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)

N-Hydroxysulfosuccinimide sodium salt is a water-soluble sulfonated analog of N-hydroxysuccinimide. It can undergo a coupling reaction with carboxylic acids in the presence of carbodiimides such as 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide(EDC) to form hydrophilic N-hydroxysulfosuccinimide active esters. These esters are useful in protein cross-linking experiments.

Formel: C₄H₄NNaO₆S
Molecular Weight: 217.13 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00043100
CAS-Nummer: 106627-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR