2-Brom-4-methylbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

565334-1G
SIAL565334-1GEA 73.6 EUR
SIAL565334-1G
2-Brom-4-methylbenzaldehyd, Sigma-Aldrich®
2-Bromo-4-methylbenzaldehyde
Formel: C₈H₇BrO
Molecular Weight: 199,05 g/mol
MDL: MFCD04039890
CAS-Nummer: 824-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR