4-Amino-2,6-dichlorpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2,6-Dichlor-4-pyridylamin, 2,6-Dichlorpyridin-4-amin

M-Clarity™ Qualitätsstufe = MQ100

565342-5G
SIAL565342-5GEA 65.9 EUR
SIAL565342-5G
4-Amino-2,6-dichlorpyridin, Sigma-Aldrich®
4-Amino-2,6-dichlorpyridin
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
MDL: MFCD00052832
CAS-Nummer: 2587-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR