4-Brom-N,N-diethylanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

565377-5G
SIAL565377-5GEA 93.7 EUR
SIAL565377-5G
4-Brom-N,N-diethylanilin, Sigma-Aldrich®
4-Brom-N,N-diethylanilin
Formel: C₁₀H₁₄BrN
Molecular Weight: 228,13 g/mol
MDL: MFCD00013530
CAS-Nummer: 2052-06-4
EINECS: 218-140-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR