3-Brom-4-methoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

565385-5G
SIAL565385-5GEA 18.8 EUR
SIAL565385-5G
3-Brom-4-methoxybenzoesäure, Sigma-Aldrich®
3-Bromo-4-methoxybenzoic acid
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
MDL: MFCD00020295
CAS-Nummer: 99-58-1
EINECS: 202-768-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR