2-Brom-4-nitroanisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromo-1-methoxy-4-nitrobenzene

M-Clarity™ Qualitätsstufe = MQ100

565407-5G
SIAL565407-5GEA 57 EUR
SIAL565407-5G
2-Brom-4-nitroanisol, Sigma-Aldrich®
2-Brom-4-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232,03 g/mol
MDL: MFCD00041242
CAS-Nummer: 5197-28-4
EINECS: 225-983-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR