Anthranilohydrazid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Aminobenzhydrazid, 2-Aminobenzoesäurehydrazid, 2-Aminobenzol-1-carbohydrazid, 2-Aminobenzohydrazid
M-Clarity™ Qualitätsstufe = MQ100
565490-10G
565490-50G
SIAL565490-10GEA
0
EUR
SIAL565490-10G
SIAL565490-50G
Anthranilohydrazid, Sigma-Aldrich®
Anthranilohydrazid
Formel:
C₇H₉N₃O Molecular Weight: 151.17 g/mol Siedepunkt: 378 Schmelzpunkt: 123 |
MDL:
MFCD00017056 CAS-Nummer: 1904-58-1 EINECS: 217-604-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...