2-Chlor-3-hydroxy-4-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-3-hydroxy-p-anisaldehyd

565997-5G
SIAL565997-5GEA 125 EUR
SIAL565997-5G
2-Chlor-3-hydroxy-4-methoxybenzaldehyd, Sigma-Aldrich®
2-Chloro-3-hydroxy-4-methoxybenzaldehyde
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
MDL: MFCD03425852
CAS-Nummer: 37687-57-3
EINECS: 253-627-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR