Bariumiodid Hydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Barium iodide dihydrate

566268-25G
SIAL566268-25GEA 108 EUR
SIAL566268-25G
Bariumiodid Hydrat, Sigma-Aldrich®
Bariumiodid Hydrat
Formel: BaI₂·H₂O
Molecular Weight: 409,15 g/mol
Dichte: 5,15 g/cm³ (20 °C)
MDL: MFCD00149155
CAS-Nummer: 7787-33-9
EINECS: 237-276-9
UN: 1564
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR