2,4-Difluorbenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Difluorbenzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

566373-5G
SIAL566373-5GEA 53 EUR
SIAL566373-5G
2,4-Difluorbenzamid, Sigma-Aldrich®
2,4-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157,12 g/mol
MDL: MFCD00015547
CAS-Nummer: 85118-02-1
EINECS: 285-654-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR