3,4-Difluorbenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Difluorbenzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

566381-5G
SIAL566381-5GEA 68.1 EUR
SIAL566381-5G
3,4-Difluorbenzamid, Sigma-Aldrich®
3,4-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157,12 g/mol
MDL: MFCD00015549
CAS-Nummer: 85118-04-3
EINECS: 285-656-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR