2-Brom-4-(trifluormethyl)phenylacetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

566438-1G
SIAL566438-1GEA 34.4 EUR
SIAL566438-1G
2-Brom-4-(trifluormethyl)phenylacetonitril, Sigma-Aldrich®
2-Brom-4-(trifluormethyl)phenylacetonitril
Formel: C₉H₅BrF₃N
Molecular Weight: 264,04 g/mol
MDL: MFCD03094308
CAS-Nummer: 474024-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR