3-(Difluormethoxy)anilin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

566446-5G
SIAL566446-5GEA 128 EUR
SIAL566446-5G
3-(Difluormethoxy)anilin, Sigma-Aldrich®
3-(Difluormethoxy)anilin
Formel: C₇H₇F₂NO
Molecular Weight: 159.14 g/mol
MDL: MFCD00236221
CAS-Nummer: 22236-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR