2-Acetylanthracen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Anthracenyl)aceton, 1-(2-Anthracenyl)ethanon, 2-Anthrylmethylketon

M-Clarity™ Qualitätsstufe = MQ100

566780-1G
SIAL566780-1GEA 100 EUR
SIAL566780-1G
2-Acetylanthracen, Sigma-Aldrich®
2-Acetylanthracen
Formel: C₁₆H₁₂O
Molecular Weight: 220,27 g/mol
MDL: MFCD00156727
CAS-Nummer: 10210-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR